RIE156322686 Rieke Metals, Inc.

2-(4-bromophenyl)-3'-cyanoacetophenone

  • MDL number MFCD02260727
  • Cas Number : 898784-14-0
  • Molecular Weight : 300.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 3-[(4-bromophenyl)acetyl]benzonitrile
Inchi Code 1S/C15H10BrNO/c16-14-6-4-11(5-7-14)9-15(18)13-3-1-2-12(8-13)10-17/h1-8H,9H2
Inchi Key PUVSXKWLTBAMHZ-UHFFFAOYSA-N

Description

Purity 97%
Country of Origin US
Product Link http://riekemetals.com/products-services/catnum/6075-2

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6075-2
Availability details for
Quantity
  • Availability
  • Add to Cart