RIE156322687 Rieke Metals, Inc.

2-(4-bromophenyl)-4'-cyanoacetophenone

  • MDL number MFCD02260728
  • Cas Number : 898784-15-1
  • Molecular Weight : 300.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6075-3

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6075-3
Availability details for
Quantity
  • Availability
  • Add to Cart