RIE156322688 Rieke Metals, Inc.

2-(2-bromophenyl)-2'-trifluoromethylacetophenone

  • MDL number MFCD02260729
  • Cas Number : 898784-16-2
  • Molecular Weight : 343.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6076-1

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6076-1
Availability details for
Quantity
  • Availability
  • Add to Cart