RIE156322689 Rieke Metals, Inc.

2-(2-bromophenyl)-3'-trifluoromethylacetophenone

  • MDL number MFCD02260730
  • Cas Number : 898784-17-3
  • Molecular Weight : 343.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6076-2

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6076-2
Availability details for
Quantity
  • Availability
  • Add to Cart