RIE156322690 Rieke Metals, Inc.

2-(2-bromophenyl)-4'-trifluoromethylacetophenone

  • MDL number MFCD02260731
  • Cas Number : 898784-18-4
  • Molecular Weight : 343.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 2-(2-bromophenyl)-1-[4-(trifluoromethyl)phenyl]ethanone
Inchi Code 1S/C15H10BrF3O/c16-13-4-2-1-3-11(13)9-14(20)10-5-7-12(8-6-10)15(17,18)19/h1-8H,9H2
Inchi Key NVRAWMKYYCBNQW-UHFFFAOYSA-N

Description

Purity 97%
Country of Origin US
Product Link http://riekemetals.com/products-services/catnum/6076-3

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6076-3
Availability details for
Quantity
  • Availability
  • Add to Cart