RIE156322691 Rieke Metals, Inc.

2-(3-bromophenyl)-2'-trifluoromethylacetophenone

  • MDL number MFCD02260732
  • Cas Number : 898784-19-5
  • Molecular Weight : 343.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 2-(3-bromophenyl)-1-[2-(trifluoromethyl)phenyl]ethanone
Inchi Code 1S/C15H10BrF3O/c16-11-5-3-4-10(8-11)9-14(20)12-6-1-2-7-13(12)15(17,18)19/h1-8H,9H2
Inchi Key WEPUGNFTQXIPJL-UHFFFAOYSA-N

Description

Purity 97%
Country of Origin US
Product Link http://riekemetals.com/products-services/catnum/6077-1

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6077-1
Availability details for
Quantity
  • Availability
  • Add to Cart