RIE156322692 Rieke Metals, Inc.

2-(3-bromophenyl)-3'-trifluoromethylacetophenone

  • MDL number MFCD02260733
  • Cas Number : 898784-20-8
  • Molecular Weight : 343.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6077-2

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6077-2
Availability details for
Quantity
  • Availability
  • Add to Cart