RIE156322699 Rieke Metals, Inc.

2-(2-Cyanophenyl)-4'-iodoacetophenone

  • MDL number MFCD02260740
  • Cas Number : 898784-33-3
  • Molecular Weight : 347.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6079-3

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6079-3
Availability details for
Quantity
  • Availability
  • Add to Cart