RIE156322702 Rieke Metals, Inc.

2-(3-Cyanophenyl)-4'-iodoacetophenone

  • MDL number MFCD02260743
  • Cas Number : 898784-39-9
  • Molecular Weight : 347.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 3-[2-(4-iodophenyl)-2-oxoethyl]benzonitrile
Inchi Code 1S/C15H10INO/c16-14-6-4-13(5-7-14)15(18)9-11-2-1-3-12(8-11)10-17/h1-8H,9H2
Inchi Key NZLFMALWMSEJAH-UHFFFAOYSA-N

Description

Purity 97%
Country of Origin US
Product Link http://riekemetals.com/products-services/catnum/6080-3

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6080-3
Availability details for
Quantity
  • Availability
  • Add to Cart