RIE156322704 Rieke Metals, Inc.

2-(4-cyanophenyl)-3'-iodoacetophenone

  • MDL number MFCD02260745
  • Cas Number : 146653-53-4
  • Molecular Weight : 347.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6081-2

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6081-2
Availability details for
Quantity
  • Availability
  • Add to Cart