RIE156322706 Rieke Metals, Inc.

2-(2-cyanophenyl)-2'-trifluoromethylacetophenone

  • MDL number MFCD02260747
  • Cas Number : 898784-45-7
  • Molecular Weight : 289.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 2-{2-oxo-2-[2-(trifluoromethyl)phenyl]ethyl}benzonitrile
Inchi Code 1S/C16H10F3NO/c17-16(18,19)14-8-4-3-7-13(14)15(21)9-11-5-1-2-6-12(11)10-20/h1-8H,9H2
Inchi Key MFZDCVQWJXWBJL-UHFFFAOYSA-N

Description

Purity 97%
Country of Origin US
Product Link http://riekemetals.com/products-services/catnum/6082-1

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6082-1
Availability details for
Quantity
  • Availability
  • Add to Cart