RIE156322707 Rieke Metals, Inc.

2-(2-cyanophenyl)-3'-trifluoromethylacetophenone

  • MDL number MFCD02260748
  • Cas Number : 898784-47-9
  • Molecular Weight : 289.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6082-2

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6082-2
Availability details for
Quantity
  • Availability
  • Add to Cart