RIE156322721 Rieke Metals, Inc.

3'-Bromo-2-(4-fluorophenyl)acetophenone

  • MDL number MFCD02260763
  • Cas Number : 898784-71-9
  • Molecular Weight : 293.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6087-2

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6087-2
Availability details for
Quantity
  • Availability
  • Add to Cart