RIE156322723 Rieke Metals, Inc.

2-(2-fluorophenyl)-2'-iodoacetophenone

  • MDL number MFCD02260765
  • Cas Number : 898784-73-1
  • Molecular Weight : 340.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6088-1

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6088-1
Availability details for
Quantity
  • Availability
  • Add to Cart