RIE156322726 Rieke Metals, Inc.

2-(3-fluorophenyl)-2'-iodoacetophenone

  • MDL number MFCD02260768
  • Cas Number : 898784-77-5
  • Molecular Weight : 340.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6089-1

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6089-1
Availability details for
Quantity
  • Availability
  • Add to Cart