RIE156322728 Rieke Metals, Inc.

2-(3-fluorophenyl)-4'-iodoacetophenone

  • MDL number MFCD02260770
  • Cas Number : 898784-81-1
  • Molecular Weight : 340.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin US
Product Link http://riekemetals.com/products-services/catnum/6089-3

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6089-3
Availability details for
Quantity
  • Availability
  • Add to Cart