RIE156322731 Rieke Metals, Inc.

2-(4-fluorophenyl)-4'-iodoacetophenone

  • MDL number MFCD02260773
  • Cas Number : 898784-87-7
  • Molecular Weight : 340.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 2-(4-fluorophenyl)-1-(4-iodophenyl)ethanone
Inchi Code 1S/C14H10FIO/c15-12-5-1-10(2-6-12)9-14(17)11-3-7-13(16)8-4-11/h1-8H,9H2
Inchi Key BKZJHBUBHFTXEL-UHFFFAOYSA-N

Description

Purity 97%
Country of Origin US
Product Link http://riekemetals.com/products-services/catnum/6090-3

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6090-3
Availability details for
Quantity
  • Availability
  • Add to Cart