RIE156322734 Rieke Metals, Inc.

4'-iodo-2-(2-methoxyphenyl)acetophenone

  • MDL number MFCD02260776
  • Cas Number : 898784-93-5
  • Molecular Weight : 352.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6091-3

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6091-3
Availability details for
Quantity
  • Availability
  • Add to Cart