RIE156322735 Rieke Metals, Inc.

1-Cyclopentyl-3-(2,4-dimethoxyphenyl)propan-1-one

  • MDL number MFCD03841033
  • Cas Number : 625445-85-4
  • Molecular Weight : 262.35

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6092

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6092
Availability details for
Quantity
  • Availability
  • Add to Cart