RIE156322738 Rieke Metals, Inc.

7-(3-oxocyclohexyl)heptanenitrile

  • MDL number MFCD03841037
  • Cas Number : 898784-99-1
  • Molecular Weight : 207.32

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6094F

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6094F
Availability details for
Quantity
  • Availability
  • Add to Cart