RIE156322742 Rieke Metals, Inc.

3-(3,5-Dimethoxybenzyl)cyclohexanone

  • MDL number MFCD03841043
  • Cas Number : 898785-03-0
  • Molecular Weight : 248.32

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6098

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6098
Availability details for
Quantity
  • Availability
  • Add to Cart