RIE156322743 Rieke Metals, Inc.

3-(2,6-Dimethylphenyl)cyclohexanone

  • MDL number MFCD03841044
  • Cas Number : 486397-33-5
  • Molecular Weight : 202.30

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6099

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6099
Availability details for
Quantity
  • Availability
  • Add to Cart