RIE156322744 Rieke Metals, Inc.

2-[(3-Oxocyclohexyl)methyl]benzonitrile

  • MDL number MFCD03841045
  • Cas Number : 898785-05-2
  • Molecular Weight : 213.28

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6100

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6100
Availability details for
Quantity
  • Availability
  • Add to Cart