RIE156322745 Rieke Metals, Inc.

3-Chloro-1-(2-methylphenyl)-1-oxopropane

  • MDL number MFCD03841046
  • Cas Number : 898785-08-5
  • Molecular Weight : 182.65

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6102B

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6102B
Availability details for
Quantity
  • Availability
  • Add to Cart