RIE156322746 Rieke Metals, Inc.

4-Chloro-1-(2-methylphenyl)-1-oxobutane

  • MDL number MFCD03841047
  • Cas Number : 40877-13-2
  • Molecular Weight : 196.68

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6102C

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6102C
Availability details for
Quantity
  • Availability
  • Add to Cart