RIE156322747 Rieke Metals, Inc.

5-chloro-1-(2-methylphenyl)-1-oxopentane

  • MDL number MFCD03841048
  • Cas Number : 898785-11-0
  • Molecular Weight : 210.71

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6102D

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6102D
Availability details for
Quantity
  • Availability
  • Add to Cart