RIE156322748 Rieke Metals, Inc.

6-chloro-1-(2-methylphenyl)-1-oxohexane

  • MDL number MFCD03841049
  • Cas Number : 898785-14-3
  • Molecular Weight : 224.73

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6102E

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6102E
Availability details for
Quantity
  • Availability
  • Add to Cart