RIE156322751 Rieke Metals, Inc.

5-chloro-1-(3-methylphenyl)-1-oxopentane

  • MDL number MFCD03841053
  • Cas Number : 898785-23-4
  • Molecular Weight : 210.71

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6103D

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6103D
Availability details for
Quantity
  • Availability
  • Add to Cart