RIE156322763 Rieke Metals, Inc.

3',3,3-trimethylbutyrophenone

  • MDL number MFCD03841075
  • Cas Number : 898785-50-7
  • Molecular Weight : 190.29

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6120-03

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6120-03
Availability details for
Quantity
  • Availability
  • Add to Cart