RIE156322765 Rieke Metals, Inc.

3,3-Dimethyl-2'-methoxybutyrophenone

  • MDL number MFCD03841077
  • Cas Number : 898764-14-2
  • Molecular Weight : 206.29

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6120-05

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6120-05
Availability details for
Quantity
  • Availability
  • Add to Cart