RIE156322767 Rieke Metals, Inc.

3,3-dimethyl-4'-methoxybutyrophenone

  • MDL number MFCD03841079
  • Cas Number : 85157-92-2
  • Molecular Weight : 206.29

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6120-07

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6120-07
Availability details for
Quantity
  • Availability
  • Add to Cart