RIE156322773 Rieke Metals, Inc.

4'-carboethoxy-3,3-dimethylbutyrophenone

  • MDL number MFCD03841085
  • Cas Number : 684271-10-1
  • Molecular Weight : 248.32

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6120-13

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6120-13
Availability details for
Quantity
  • Availability
  • Add to Cart