RIE156322774 Rieke Metals, Inc.

3,3-dimethyl-2'-thiomethylbutyrophenone

  • MDL number MFCD07699543
  • Cas Number : 898764-32-4
  • Molecular Weight : 222.35

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 3,3-dimethyl-1-[2-(methylsulfanyl)phenyl]-1-butanone
Inchi Code 1S/C13H18OS/c1-13(2,3)9-11(14)10-7-5-6-8-12(10)15-4/h5-8H,9H2,1-4H3
Inchi Key IZCPSCQROHEDCO-UHFFFAOYSA-N

Description

Purity 97%
Country of Origin US
Product Link http://riekemetals.com/products-services/catnum/6120-14

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6120-14
Availability details for
Quantity
  • Availability
  • Add to Cart