RIE156322775 Rieke Metals, Inc.

3,3-dimethyl-4'-thiomethylbutyrophenone

  • MDL number MFCD07699544
  • Cas Number : 440121-15-3
  • Molecular Weight : 222.35

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6120-15

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6120-15
Availability details for
Quantity
  • Availability
  • Add to Cart