RIE156322777 Rieke Metals, Inc.

4'-Bromo-3,3-dimethylbutyrophenone

  • MDL number MFCD03841089
  • Cas Number : 898764-38-0
  • Molecular Weight : 255.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 1-(4-bromophenyl)-3,3-dimethyl-1-butanone
Inchi Code 1S/C12H15BrO/c1-12(2,3)8-11(14)9-4-6-10(13)7-5-9/h4-7H,8H2,1-3H3
Inchi Key RMSYXFAVKLWJDS-UHFFFAOYSA-N

Description

Purity 97%
Country of Origin US
Product Link http://riekemetals.com/products-services/catnum/6120-19

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6120-19
Availability details for
Quantity
  • Availability
  • Add to Cart