RIE156322783 Rieke Metals, Inc.

2',3,3,4'-tetramethylbutyrophenone

  • MDL number MFCD03841094
  • Cas Number : 898764-47-1
  • Molecular Weight : 204.31

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6120-27

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6120-27
Availability details for
Quantity
  • Availability
  • Add to Cart