RIE156322786 Rieke Metals, Inc.

3,3,3',4'-tetramethylbutyrophenone

  • MDL number MFCD03841097
  • Cas Number : 898764-56-2
  • Molecular Weight : 204.31

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6120-30

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6120-30
Availability details for
Quantity
  • Availability
  • Add to Cart