RIE156322788 Rieke Metals, Inc.

4'-bromo-3,3-dimethyl-3'-fluorobutyrophenone

  • MDL number MFCD07699546
  • Cas Number : 898764-62-0
  • Molecular Weight : 273.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6120-32

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6120-32
Availability details for
Quantity
  • Availability
  • Add to Cart