RIE156322791 Rieke Metals, Inc.

2'-Chloro-3,3-dimethylbutyrophenone

  • MDL number MFCD07699549
  • Cas Number : 898764-68-6
  • Molecular Weight : 210.71

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6120-36

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6120-36
Availability details for
Quantity
  • Availability
  • Add to Cart