RIE156322796 Rieke Metals, Inc.

4'-Bromo-3,3-dimethyl-2'-fluorobutyrophenone

  • MDL number MFCD03841103
  • Cas Number : 898764-78-8
  • Molecular Weight : 273.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6120-49

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6120-49
Availability details for
Quantity
  • Availability
  • Add to Cart