RIE156322797 Rieke Metals, Inc.

3'-chloro-3,3-dimethyl-5'-fluorobutyrophenone

  • MDL number MFCD03841105
  • Cas Number : 898764-80-2
  • Molecular Weight : 228.70

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6120-51

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6120-51
Availability details for
Quantity
  • Availability
  • Add to Cart