RIE156322798 Rieke Metals, Inc.

4'-chloro-3,3-dimethyl-2'-fluorobutyrophenone

  • MDL number MFCD03841106
  • Cas Number : 898764-82-4
  • Molecular Weight : 228.70

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6120-52

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6120-52
Availability details for
Quantity
  • Availability
  • Add to Cart