RIE156322801 Rieke Metals, Inc.

2',5'-dichloro-3,3-dimethylbutyrophenone

  • MDL number MFCD03841109
  • Cas Number : 898764-88-0
  • Molecular Weight : 245.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6120-55

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6120-55
Availability details for
Quantity
  • Availability
  • Add to Cart