RIE156322803 Rieke Metals, Inc.

3',5'-Dichloro-3,3-dimethylbutyrophenone

  • MDL number MFCD03841111
  • Cas Number : 898764-92-6
  • Molecular Weight : 245.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6120-57

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6120-57
Availability details for
Quantity
  • Availability
  • Add to Cart