RIE156322807 Rieke Metals, Inc.

3,3-dimethyl-3',4',5'-trifluorobutyrophenone

  • MDL number MFCD03841115
  • Cas Number : 898765-00-9
  • Molecular Weight : 230.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6120-61

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6120-61
Availability details for
Quantity
  • Availability
  • Add to Cart