RIE156322809 Rieke Metals, Inc.

3',2,2-Trimethylbutyrophenone

  • MDL number MFCD03841117
  • Cas Number : 898765-02-1
  • Molecular Weight : 190.29

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6121-03

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6121-03
Availability details for
Quantity
  • Availability
  • Add to Cart