RIE156322811 Rieke Metals, Inc.

2,2-dimethyl-2'-methoxybutyrophenone

  • MDL number MFCD03841119
  • Cas Number : 898765-04-3
  • Molecular Weight : 206.29

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6121-05

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6121-05
Availability details for
Quantity
  • Availability
  • Add to Cart