RIE156322818 Rieke Metals, Inc.

3'-carboethoxy-2,2-dimethylbutyrophenone

  • MDL number MFCD03841126
  • Cas Number : 898765-22-5
  • Molecular Weight : 248.32

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6121-12

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6121-12
Availability details for
Quantity
  • Availability
  • Add to Cart