RIE156322819 Rieke Metals, Inc.

4'-carboethoxy-2,2-dimethylbutyrophenone

  • MDL number MFCD03841127
  • Cas Number : 898765-25-8
  • Molecular Weight : 248.32

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6121-13

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6121-13
Availability details for
Quantity
  • Availability
  • Add to Cart