RIE156322820 Rieke Metals, Inc.

2,2-dimethyl-2'-thiomethylbutyrophenone

  • MDL number MFCD03841128
  • Cas Number : 898765-28-1
  • Molecular Weight : 222.35

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6121-14

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6121-14
Availability details for
Quantity
  • Availability
  • Add to Cart